| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006373.hg.1 |
ENST00000225927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006373.hg.1 |
ENST00000377405.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006373.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006373.hg.1 |
TC17000526.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000592454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
ENST00000592454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
TC1700012259.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202365.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
TC1700012259.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700202366.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000225927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000586516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000590358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000591587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000592454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
ENST00000592454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012259.hg.1 |
NM_000263.3 |
Refseq |