Assay Details
| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297436.1 | NP_001284365.1 | 1 - 2 | 69 | 99 | |
| GenBank mRNA | AM904814.1 | - | 1 - 2 | 71 | 99 | |
| BC035837.1 | - | 1 - 2 | 51 | 5589083 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AM904814.1, BC035837.1, NM_001297436.1 transcripts.
Set Membership: |
|

HAS1
Genomic Map