Assay Details
| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278282.1 | NP_001265211.1 | 1 - 2 | 218 | 82 | |
| NM_001318931.1 | NP_001305860.1 | 1 - 2 | 218 | 82 | ||
| GenBank mRNA | DA336224.1 | - | 1 - 2 | 127 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278282.1,NM_001318931.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

RPE
Genomic Map