Assay Details
Gene Symbol: SIRPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178460.2 | NP_848555.2 | 1 - 2 | 119 | 130 | |
GenBank mRNA | AK093083.1 | - | 1 - 2 | 1265 | 130 | |
BC033502.1 | - | 1 - 2 | 119 | 5172531 | 130 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000553.hg
Additional Information:
This assay is in the 5'UTR region of the AK093083.1,BC033502.1,NM_178460.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20008023.hg.1 | ENST00000381621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008023.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008023.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008023.hg.1 | TC20000553.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20008025.hg.1 | ENST00000381621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008025.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008025.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008025.hg.1 | TC20000553.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20008024.hg.1 | ENST00000381621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008024.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008024.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008024.hg.1 | TC20000553.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | TC2000009969.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170871.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | TC2000009969.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170875.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000429387.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009969.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | TC2000009969.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | ENST00000453770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170872.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | NM_178460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | TC2000009969.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | ENST00000381621.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | ENST00000381623.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170876.hg.1 | NM_178460.2 | Refseq |
