Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282488.1 | NP_001269417.1 | 1 - 2 | 121 | 110 | |
| GenBank mRNA | BI547222.1 | - | 1 - 2 | 126 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001282488.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
telomere maintenance via recombination
DNA replication, synthesis of RNA primer
DNA replication initiation
DNA strand elongation involved in DNA replication
DNA binding
DNA primase activity
metal ion binding
4 iron, 4 sulfur cluster binding
G1/S transition of mitotic cell cycle
telomere maintenance via recombination
DNA replication, synthesis of RNA primer
DNA replication initiation
DNA strand elongation involved in DNA replication
DNA binding
DNA primase activity
metal ion binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000490313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000490313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000490313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000550475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000550475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000550475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198918.hg.1 | TC0600014139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198917.hg.1 | TC0600014139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000274891.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000419977.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000470638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000490313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000490313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000490313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000550475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000550475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000550475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | ENST00000615550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_000947.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014139.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198911.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198911.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198911.hg.1 | ENST00000370687.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198911.hg.1 | NM_001282488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198911.hg.1 | TC0600014139.hg.1 | Clariom D TC |

PRIM2
Genomic Map