Assay Details
| Gene Symbol: HENMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102592.1 | NP_001096062.1 | 6 - 7 | 815 | 125 | |
| NM_144584.2 | NP_653185.2 | 6 - 7 | 998 | 125 | ||
| XM_005270411.1 | XP_005270468.1 | 5 - 6 | 697 | 125 | ||
| XM_017000179.1 | XP_016855668.1 | 5 - 6 | 799 | 125 | ||
| GenBank mRNA | AK055087.1 | - | 6 - 7 | 998 | 125 | |
| AK310152.1 | - | 6 - 7 | 988 | 125 | ||
| BC012198.1 | - | 6 - 7 | 724 | 2906200 | 125 | |
| BC088366.1 | - | 6 - 7 | 735 | 30390911 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002945.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | ENST00000370031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | ENST00000370032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | ENST00000402983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045763.hg.1 | TC01002945.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212996.hg.1 | TC0100015171.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000370032.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000402983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000420055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000483729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | ENST00000493676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_001102592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015171.hg.1 | NM_144584.2 | Refseq |

HENMT1
Genomic Map