Assay Details
| Gene Symbol: NAGLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000263.3 | NP_000254.2 | 1 - 2 | 722 | 137 | |
| XM_006721920.2 | XP_006721983.1 | 1 - 2 | 115 | 137 | ||
| XM_011524840.1 | XP_011523142.1 | 1 - 2 | 115 | 137 | ||
| GenBank mRNA | BC053991.1 | - | 1 - 2 | 405 | 6213852 | 137 |
| L78464.1 | - | 1 - 2 | 388 | 137 | ||
| U40846.1 | - | 1 - 2 | 483 | 137 | ||
| U43573.1 | - | 1 - 2 | 382 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000526.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_006721920.2,XM_011524840.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17006372.hg.1 | ENST00000225927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006372.hg.1 | ENST00000377405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006372.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006372.hg.1 | TC17000526.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006370.hg.1 | ENST00000225927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006370.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006370.hg.1 | TC17000526.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000592454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | ENST00000592454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006371.hg.1 | ENST00000225927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006371.hg.1 | ENST00000377405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006371.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006371.hg.1 | TC17000526.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000592454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | ENST00000592454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012259.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202359.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202359.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202359.hg.1 | TC1700012259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202359.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202359.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202357.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202357.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202357.hg.1 | TC1700012259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202357.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202357.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | TC1700012259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | ENST00000586516.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | ENST00000591587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202361.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | TC1700012259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | ENST00000225927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | ENST00000590358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202363.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202356.hg.1 | NM_000263.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202356.hg.1 | TC1700012259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202356.hg.1 | NM_000263.3 | Refseq |

NAGLU
Genomic Map