Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014157.3 | NP_054876.2 | 2 - 3 | 313 | 87 | |
| XM_011523038.2 | XP_011521340.1 | 2 - 3 | 168 | 87 | ||
| XM_011523039.2 | XP_011521341.1 | 4 - 5 | 1061 | 87 | ||
| XM_011523040.2 | XP_011521342.1 | 2 - 3 | 586 | 87 | ||
| XM_011523041.2 | XP_011521343.1 | 2 - 3 | 154 | 87 | ||
| XM_011523042.2 | XP_011521344.1 | 2 - 3 | 313 | 87 | ||
| XM_017023176.1 | XP_016878665.1 | 3 - 4 | 476 | 87 | ||
| GenBank mRNA | AF161550.1 | - | 2 - 3 | 272 | 87 | |
| AK314300.1 | - | 2 - 3 | 302 | 87 | ||
| AL136785.1 | - | 2 - 3 | 290 | 87 | ||
| BC126445.1 | - | 2 - 3 | 282 | 8992161 | 87 | |
| BC126447.1 | - | 2 - 3 | 282 | 8992163 | 87 | |
| CR533506.1 | - | 2 - 3 | 234 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000501.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008028.hg.1 | ENST00000561923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008028.hg.1 | ENST00000561923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007029.hg.1 | ENST00000219299.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007029.hg.1 | TC16000501.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000443128.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000561517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000566498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_001142302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008029.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | TC1600008029.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | ENST00000219299.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | ENST00000569374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | ENST00000616795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159245.hg.1 | NM_014157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008028.hg.1 | ENST00000561923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008028.hg.1 | ENST00000561923.1 | Ensembl |

CCDC113
Genomic Map