| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007032.hg.1 |
ENST00000219299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007032.hg.1 |
ENST00000443128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007032.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007032.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007032.hg.1 |
TC16000501.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000566498.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000569374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008029.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
NM_014157.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
TC1600008029.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000219299.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000443128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000561517.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
ENST00000616795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
NM_001142302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159249.hg.1 |
NM_014157.3 |
Refseq |