Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008840.hg.1 |
TC05000639.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008840.hg.1 |
ENST00000265335.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008840.hg.1 |
ENST00000378823.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008840.hg.1 |
NM_005732.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000378823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000423956.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000453394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000533482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
NM_005732.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000378823.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000423956.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000453394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
ENST00000533482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
NM_005732.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159259.hg.1 |
TC0500008641.hg.1 |
Clariom D TC |