Assay Details
| Gene Symbol: DENND6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152678.2 | NP_689891.1 | 14 - 15 | 1328 | 116 | |
| XM_006713019.3 | XP_006713082.1 | 14 - 15 | 1354 | 116 | ||
| XM_006713020.2 | XP_006713083.1 | 15 - 16 | 1560 | 116 | ||
| XM_017005864.1 | XP_016861353.1 | 15 - 16 | 1563 | 116 | ||
| GenBank mRNA | AK074156.1 | - | 14 - 15 | 1315 | 116 | |
| AK092288.1 | - | 9 - 10 | 730 | 116 | ||
| BC040291.1 | - | 14 - 15 | 1316 | 4822560 | 116 | |
| BC054343.1 | - | 5 - 6 | 411 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001496.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03027255.hg.1 | ENST00000311128.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027255.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027255.hg.1 | TC03001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027253.hg.1 | ENST00000311128.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027253.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027253.hg.1 | TC03001496.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184618.hg.1 | TC0300011276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184616.hg.1 | TC0300011276.hg.1 | Clariom D TC |

DENND6A
Genomic Map