Assay Details
| Gene Symbol: DENND6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152678.2 | NP_689891.1 | 7 - 8 | 773 | 75 | |
| XM_006713019.3 | XP_006713082.1 | 7 - 8 | 799 | 75 | ||
| XM_006713020.2 | XP_006713083.1 | 8 - 9 | 1005 | 75 | ||
| XM_017005864.1 | XP_016861353.1 | 8 - 9 | 1008 | 75 | ||
| GenBank mRNA | AK074156.1 | - | 7 - 8 | 760 | 75 | |
| AK092288.1 | - | 2 - 3 | 175 | 75 | ||
| BC040291.1 | - | 7 - 8 | 761 | 4822560 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001496.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK092288.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03027264.hg.1 | ENST00000311128.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027264.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027264.hg.1 | TC03001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184629.hg.1 | TC0300011276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000311128.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000464875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000471531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000477344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000480710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | ENST00000487662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011276.hg.1 | NM_152678.2 | Refseq |

DENND6A
Genomic Map