Assay Details
| Gene Symbol: GPR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC032694.1 | - | 2 - 3 | 1538 | 5228678 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000438398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000438398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000438398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000438398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000438398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000438398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016000.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217646.hg.1 | TC0200016000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000392039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217650.hg.1 | TC0200016000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217647.hg.1 | TC0200016000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | ENST00000392040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | ENST00000444078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | ENST00000622008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | NM_005683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217649.hg.1 | TC0200016000.hg.1 | Clariom D TC |

GPR55
Genomic Map