Assay Details
| Gene Symbol: SPATA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139073.3 | NP_620712.2 | 3 - 4 | 725 | 93 | |
| GenBank mRNA | AK093385.1 | - | 3 - 4 | 723 | 93 | |
| AY032925.1 | - | 4 - 5 | 823 | 93 | ||
| BC047704.1 | - | 4 - 5 | 668 | 5744971 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001384.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | ENST00000355662.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | ENST00000424440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | ENST00000433428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | ENST00000452881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021710.hg.1 | TC02001384.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021711.hg.1 | ENST00000355662.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021711.hg.1 | ENST00000424440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021711.hg.1 | ENST00000433428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021711.hg.1 | ENST00000452881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021711.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021711.hg.1 | TC02001384.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179261.hg.1 | TC0200011048.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179260.hg.1 | TC0200011048.hg.1 | Clariom D TC |

SPATA3
Genomic Map