Assay Details
| Gene Symbol: SUSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019601.3 | NP_062547.1 | 8 - 9 | 1389 | 76 | |
| GenBank mRNA | AK026431.1 | - | 8 - 9 | 1407 | 76 | |
| AK126105.1 | - | 6 - 7 | 2238 | 76 | ||
| BC033107.1 | - | 8 - 9 | 1389 | 5457186 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000146.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002685.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002685.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002685.hg.1 | TC22000146.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006883.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | NM_019601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | TC2200006883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | ENST00000358321.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | ENST00000463101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147599.hg.1 | NM_019601.3 | Refseq |

SUSD2
Genomic Map