| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013377.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013377.hg.1 |
TC09001014.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013375.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013375.hg.1 |
TC09001014.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013374.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013374.hg.1 |
TC09001014.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013376.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013376.hg.1 |
TC09001014.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900167398.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167398.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167398.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167398.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167398.hg.1 |
TC0900009854.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900167384.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167384.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167384.hg.1 |
TC0900009854.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900167394.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167394.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167394.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167394.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167394.hg.1 |
TC0900009854.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900167397.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167397.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167397.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167397.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167397.hg.1 |
TC0900009854.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900167400.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167400.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167400.hg.1 |
TC0900009854.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009854.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167408.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167408.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167408.hg.1 |
ENST00000379514.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167408.hg.1 |
NR_003667.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167408.hg.1 |
TC0900009854.hg.1 |
Clariom D TC |