Assay Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014990.2 | NP_001014990.1 | 4 - 5 | 260 | 100 | |
| NM_014180.3 | NP_054899.2 | 5 - 6 | 378 | 100 | ||
| GenBank mRNA | AF161507.1 | - | 5 - 6 | 346 | 100 | |
| AK000601.1 | - | 5 - 6 | 365 | 100 | ||
| AK097880.1 | - | 1 - 2 | 2015 | 100 | ||
| AK302784.1 | - | 4 - 5 | 240 | 100 | ||
| BC012565.1 | - | 5 - 6 | 348 | 4333117 | 100 | |
| BM662504.1 | - | 2 - 3 | 100 | |||
| BP372420.1 | - | 5 - 6 | 378 | 100 | ||
| BU943770.1 | - | 5 - 6 | 371 | 100 | ||
| CR933679.1 | - | 4 - 5 | 239 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000857.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | ENST00000265229.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | ENST00000439747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | ENST00000522038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | ENST00000523037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012521.hg.1 | TC05000857.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164317.hg.1 | TC0500009183.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000265229.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000439747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000518364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000519059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000520040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000522038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000523037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | ENST00000614620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_001014990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009183.hg.1 | NM_014180.3 | Refseq |

MRPL22
Genomic Map