Assay Details
| Gene Symbol: PLPPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024888.2 | NP_079164.1 | 5 - 6 | 663 | 125 | |
| XM_011528317.2 | XP_011526619.1 | 5 - 6 | 1255 | 125 | ||
| GenBank mRNA | AF541282.1 | - | 5 - 6 | 663 | 125 | |
| AL136596.1 | - | 5 - 6 | 663 | 125 | ||
| AY304517.1 | - | 5 - 6 | 663 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000990.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19013503.hg.1 | ENST00000300947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013503.hg.1 | ENST00000359894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013503.hg.1 | ENST00000520876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013503.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013503.hg.1 | TC19000990.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013501.hg.1 | ENST00000300947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013501.hg.1 | ENST00000359894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013501.hg.1 | ENST00000520876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013501.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013501.hg.1 | TC19000990.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013502.hg.1 | ENST00000359894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013502.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013502.hg.1 | TC19000990.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013504.hg.1 | ENST00000300947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013504.hg.1 | ENST00000359894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013504.hg.1 | ENST00000520876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013504.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013504.hg.1 | TC19000990.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000519502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009112.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | TC1900009112.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172344.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172342.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172342.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172342.hg.1 | TC1900009112.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172342.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172342.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | NM_024888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | TC1900009112.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | ENST00000359894.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | ENST00000517665.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | ENST00000520876.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | NM_001270366.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172343.hg.1 | NM_024888.2 | Refseq |

PLPPR3
Genomic Map