Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053963.hg.1 |
TC01003466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053963.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053963.hg.1 |
ENST00000538148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053963.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000384263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000384263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000461664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000463610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000471457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
ENST00000491436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016260.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223775.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223775.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223775.hg.1 |
ENST00000354775.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223775.hg.1 |
NM_000696.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223775.hg.1 |
TC0100016260.hg.1 |
Clariom D TC |