Assay Details
| Gene Symbol: ANKRD13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030816.4 | NP_110443.3 | 11 - 12 | 80 | ||
| XM_006710929.3 | XP_006710992.1 | 11 - 12 | 80 | |||
| XM_017002413.1 | XP_016857902.1 | 9 - 10 | 80 | |||
| XM_017002414.1 | XP_016857903.1 | 10 - 11 | 80 | |||
| XM_017002415.1 | XP_016857904.1 | 10 - 11 | 80 | |||
| XM_017002416.1 | XP_016857905.1 | 6 - 7 | 80 | |||
| GenBank mRNA | AK074187.1 | - | 3 - 4 | 80 | ||
| AL136717.1 | - | 10 - 11 | 80 | |||
| BC028840.1 | - | 11 - 12 | 4558748 | 80 | ||
| BU622247.1 | - | 2 - 3 | 286 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002771.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01043272.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043272.hg.1 | ENST00000370944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043272.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043272.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043272.hg.1 | TC01002771.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208721.hg.1 | TC0100014566.hg.1 | Clariom D TC |

ANKRD13C
Genomic Map