Assay Details
| Gene Symbol: ANKRD13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030816.4 | NP_110443.3 | 4 - 5 | 995 | 101 | |
| XM_005271234.3 | XP_005271291.1 | 4 - 5 | 1238 | 101 | ||
| XM_005271235.3 | XP_005271292.1 | 4 - 5 | 985 | 101 | ||
| XM_006710929.3 | XP_006710992.1 | 4 - 5 | 1008 | 101 | ||
| XM_017002413.1 | XP_016857902.1 | 2 - 3 | 218 | 101 | ||
| XM_017002415.1 | XP_016857904.1 | 4 - 5 | 346 | 101 | ||
| GenBank mRNA | AL136717.1 | - | 3 - 4 | 891 | 101 | |
| BC028840.1 | - | 4 - 5 | 984 | 4558748 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002771.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017002415.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043282.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043282.hg.1 | ENST00000370944.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043282.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043282.hg.1 | TC01002771.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000464236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | ENST00000498735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014566.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | ENST00000262346.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | ENST00000370944.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | ENST00000490846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | NM_030816.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208737.hg.1 | TC0100014566.hg.1 | Clariom D TC |

ANKRD13C
Genomic Map