Assay Details
| Gene Symbol: OPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033282.3 | NP_150598.1 | 2 - 3 | 512 | 81 | |
| XM_017016956.1 | XP_016872445.1 | 2 - 3 | 1574 | 81 | ||
| GenBank mRNA | BC113558.1 | - | 2 - 3 | 341 | 8322610 | 81 |
| CD634966.1 | - | 2 - 3 | 81 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002926.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006582.hg.1 | ENST00000241891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006582.hg.1 | ENST00000372071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006582.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006582.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006582.hg.1 | TC10002926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156290.hg.1 | TC1000008309.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |

OPN4
Genomic Map