Assay Details
| Gene Symbol: OPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030015.2 | NP_001025186.1 | 4 - 5 | 687 | 62 | |
| NM_033282.3 | NP_150598.1 | 3 - 4 | 654 | 62 | ||
| XM_017016955.1 | XP_016872444.1 | 4 - 5 | 1749 | 62 | ||
| XM_017016956.1 | XP_016872445.1 | 3 - 4 | 1716 | 62 | ||
| XM_017016957.1 | XP_016872446.1 | 4 - 5 | 1484 | 62 | ||
| GenBank mRNA | BC029291.1 | - | 4 - 5 | 627 | 62 | |
| BC113558.1 | - | 3 - 4 | 483 | 8322610 | 62 | |
| BC143688.1 | - | 4 - 5 | 516 | 9052202 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002926.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006583.hg.1 | ENST00000241891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006583.hg.1 | ENST00000372071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006583.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006583.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006583.hg.1 | TC10002926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156291.hg.1 | TC1000008309.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |

OPN4
Genomic Map