Assay Details
| Gene Symbol: OPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030015.2 | NP_001025186.1 | 9 - 10 | 1513 | 88 | |
| NM_033282.3 | NP_150598.1 | 8 - 9 | 1480 | 88 | ||
| XM_017016955.1 | XP_016872444.1 | 9 - 10 | 2575 | 88 | ||
| XM_017016956.1 | XP_016872445.1 | 8 - 9 | 2542 | 88 | ||
| XM_017016957.1 | XP_016872446.1 | 9 - 10 | 2310 | 88 | ||
| GenBank mRNA | AK310520.1 | - | 3 - 4 | 431 | 88 | |
| BC029291.1 | - | 9 - 10 | 1453 | 88 | ||
| BC113558.1 | - | 8 - 9 | 1309 | 8322610 | 88 | |
| BC143688.1 | - | 9 - 10 | 1342 | 9052202 | 88 | |
| KT381095.1 | - | 2 - 3 | 239 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002926.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006588.hg.1 | ENST00000241891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006588.hg.1 | ENST00000372071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006588.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006588.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006588.hg.1 | TC10002926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006589.hg.1 | ENST00000241891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006589.hg.1 | ENST00000372071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006589.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006589.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006589.hg.1 | TC10002926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006587.hg.1 | ENST00000241891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006587.hg.1 | ENST00000372071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006587.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006587.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006587.hg.1 | TC10002926.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008309.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156296.hg.1 | TC1000008309.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156294.hg.1 | TC1000008309.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | ENST00000241891.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | ENST00000372071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | ENST00000443292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | NM_001030015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | NM_033282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156295.hg.1 | TC1000008309.hg.1 | Clariom D TC |

OPN4
Genomic Map