| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018755.hg.1 |
ENST00000372027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018755.hg.1 |
ENST00000474994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018755.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018755.hg.1 |
TC10001482.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000488950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000488950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187464.hg.1 |
TC1000012573.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000372027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000488950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000488950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000608753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
ENST00000610081.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012573.hg.1 |
NM_024756.2 |
Refseq |