Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024756.2 | NP_079032.2 | 5 - 6 | 785 | 82 | |
| XM_005270153.1 | XP_005270210.1 | 5 - 6 | 824 | 82 | ||
| XM_006717970.3 | XP_006718033.1 | 5 - 6 | 2764 | 82 | ||
| GenBank mRNA | AK023527.1 | - | 5 - 6 | 732 | 82 | |
| AK304727.1 | - | 5 - 6 | 642 | 82 | ||
| AK308401.1 | - | 4 - 5 | 626 | 82 | ||
| BC064415.1 | - | 5 - 6 | 740 | 82 | ||
| BC072456.1 | - | 5 - 6 | 681 | 82 | ||
| BC094744.1 | - | 5 - 6 | 732 | 30343159 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001482.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000488950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000488950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018751.hg.1 | ENST00000372027.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018751.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018751.hg.1 | TC10001482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018744.hg.1 | ENST00000372027.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018744.hg.1 | ENST00000443699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018744.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018744.hg.1 | TC10001482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018750.hg.1 | ENST00000372027.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018750.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018750.hg.1 | TC10001482.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000488950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000488950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000608753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012573.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187459.hg.1 | TC1000012573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000187458.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187458.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187458.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187458.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187458.hg.1 | TC1000012573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | ENST00000488950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | ENST00000372027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | ENST00000488950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | ENST00000610081.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | NM_024756.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187460.hg.1 | TC1000012573.hg.1 | Clariom D TC |

MMRN2
Genomic Map