| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014064.hg.1 |
ENST00000376070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014064.hg.1 |
ENST00000376087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014064.hg.1 |
ENST00000436985.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014064.hg.1 |
NM_001256053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014064.hg.1 |
NM_014915.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014064.hg.1 |
TC10001124.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
ENST00000376087.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
ENST00000436985.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
NM_001256053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
NM_014915.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
ENST00000376087.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
ENST00000436985.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
NM_001256053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
NM_014915.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000168876.hg.1 |
TC1000010113.hg.1 |
Clariom D TC |