Assay Details
| Gene Symbol: CSN1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025104.1 | NP_001020275.1 | 2 - 3 | 100 | 63 | |
| NM_001890.1 | NP_001881.1 | 2 - 3 | 100 | 63 | ||
| XM_006714089.2 | XP_006714152.1 | 2 - 3 | 119 | 63 | ||
| XM_006714090.2 | XP_006714153.1 | 2 - 3 | 119 | 63 | ||
| XM_006714091.2 | XP_006714154.1 | 2 - 3 | 119 | 63 | ||
| GenBank mRNA | BC128227.1 | - | 2 - 3 | 89 | 40114618 | 63 |
| BC128228.1 | - | 2 - 3 | 89 | 40114622 | 63 | |
| BG217918.1 | - | 1 - 2 | 62 | 63 | ||
| DQ064604.1 | - | 1 - 2 | 51 | 63 | ||
| U23157.1 | - | 2 - 3 | 79 | 63 | ||
| X78416.1 | - | 2 - 3 | 100 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000374.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000633258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000633258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | ENST00000633258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | ENST00000246891.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | ENST00000354865.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | ENST00000444405.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | ENST00000505782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | ENST00000507763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | ENST00000507772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005060.hg.1 | TC04000374.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000510936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000633258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000633258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | ENST00000633258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007769.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000246891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000505782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000507763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | ENST00000507772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | NM_001025104.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | NM_001890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152904.hg.1 | TC0400007769.hg.1 | Clariom D TC |

CSN1S1
Genomic Map