Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 28 - 29 | 3224 | 71 | |
| XM_011538537.2 | XP_011536839.1 | 28 - 29 | 3265 | 71 | ||
| XM_011538538.2 | XP_011536840.1 | 28 - 29 | 3265 | 71 | ||
| XM_011538539.2 | XP_011536841.1 | 14 - 15 | 1468 | 71 | ||
| XM_011538541.2 | XP_011536843.1 | 6 - 7 | 575 | 71 | ||
| XM_017019585.1 | XP_016875074.1 | 28 - 29 | 3265 | 71 | ||
| GenBank mRNA | AB052958.1 | - | 28 - 29 | 3220 | 71 | |
| AJ295695.1 | - | 28 - 29 | 3224 | 71 | ||
| AK074051.1 | - | 6 - 7 | 595 | 71 | ||
| AK160380.1 | - | 27 - 28 | 2937 | 71 | ||
| AY311388.1 | - | 28 - 29 | 3242 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Set Membership: |
|

STAB2
Genomic Map