| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001399.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001399.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001399.hg.1 |
TC21000160.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001402.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001402.hg.1 |
ENST00000485272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001402.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001402.hg.1 |
TC21000160.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001401.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001401.hg.1 |
ENST00000485272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001401.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001401.hg.1 |
TC21000160.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001400.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001400.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21001400.hg.1 |
TC21000160.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
TC2100007066.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147721.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
TC2100007066.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147722.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000485272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
ENST00000490393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007066.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147718.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147718.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147718.hg.1 |
TC2100007066.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100147718.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147718.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147720.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147720.hg.1 |
NM_018962.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100147720.hg.1 |
TC2100007066.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100147720.hg.1 |
ENST00000329553.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100147720.hg.1 |
NM_018962.2 |
Refseq |