Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080824.2 | NP_001074293.1 | 3 - 4 | 1051 | 76 | |
| NM_001277053.1 | NP_001263982.1 | 4 - 5 | 1198 | 76 | ||
| XM_011532504.1 | XP_011530806.1 | 4 - 5 | 998 | 76 | ||
| GenBank mRNA | BC051789.1 | - | 3 - 4 | 1052 | 76 | |
| JQ653415.1 | - | 4 - 5 | 992 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002034.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02032497.hg.1 | ENST00000335459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032497.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032497.hg.1 | ENST00000409520.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032497.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032497.hg.1 | TC02002034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000436322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000436322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032492.hg.1 | ENST00000335459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032492.hg.1 | ENST00000409520.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032492.hg.1 | ENST00000479944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032492.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032492.hg.1 | TC02002034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032491.hg.1 | ENST00000335459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032491.hg.1 | ENST00000409520.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032491.hg.1 | ENST00000479944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032491.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02032491.hg.1 | TC02002034.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000436322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000436322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000460991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000495990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016699.hg.1 | NM_001307978.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000335459.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000409520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000479944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | ENST00000496500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | NM_001080824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | NM_001277053.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227043.hg.1 | TC0200016699.hg.1 | Clariom D TC |

TRABD2A
Genomic Map