Assay Details
| Gene Symbol: RRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003683.5 | NP_003674.1 | 6 - 7 | 646 | 123 | |
| XM_017028485.1 | XP_016883974.1 | 6 - 7 | 637 | 123 | ||
| XM_017028486.1 | XP_016883975.1 | 6 - 7 | 628 | 123 | ||
| GenBank mRNA | AK223185.1 | - | 6 - 7 | 607 | 123 | |
| AK223215.1 | - | 6 - 7 | 603 | 123 | ||
| AK303000.1 | - | 6 - 7 | 606 | 123 | ||
| AK313116.1 | - | 6 - 7 | 603 | 123 | ||
| AL137757.1 | - | 3 - 4 | 667 | 123 | ||
| AY033999.1 | - | 1 - 2 | 192 | 123 | ||
| BC000380.2 | - | 6 - 7 | 593 | 2820455 | 123 | |
| BC014787.1 | - | 6 - 7 | 568 | 123 | ||
| U79775.1 | - | 6 - 7 | 591 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000214.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002350.hg.1 | ENST00000400387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002350.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002350.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002350.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002350.hg.1 | TC21000214.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | NM_003683.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | TC2100007327.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000467112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000471909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000483896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | ENST00000497547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100149414.hg.1 | NM_003683.5 | Refseq |

RRP1
Genomic Map