Assay Details
Gene Symbol: RRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003683.5 | NP_003674.1 | 8 - 9 | 904 | 73 | |
XM_017028485.1 | XP_016883974.1 | 8 - 9 | 895 | 73 | ||
XM_017028486.1 | XP_016883975.1 | 8 - 9 | 886 | 73 | ||
GenBank mRNA | AK223185.1 | - | 8 - 9 | 865 | 73 | |
AK223215.1 | - | 8 - 9 | 861 | 73 | ||
AK303000.1 | - | 8 - 9 | 864 | 73 | ||
AL137757.1 | - | 5 - 6 | 925 | 73 | ||
AY033999.1 | - | 3 - 4 | 450 | 73 | ||
BC000380.2 | - | 8 - 9 | 851 | 2820455 | 73 | |
BC014787.1 | - | 8 - 9 | 826 | 73 | ||
BI223794.1 | - | 3 - 4 | 278 | 73 | ||
U79775.1 | - | 8 - 9 | 849 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000214.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR21002354.hg.1 | ENST00000400387.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002354.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002354.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002354.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002354.hg.1 | TC21000214.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21002353.hg.1 | ENST00000400387.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002353.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002353.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002353.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002353.hg.1 | TC21000214.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | TC2100007327.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149417.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | TC2100007327.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100149418.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000467112.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000471909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000473988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000475534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000483896.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000492638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | ENST00000497547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007327.hg.1 | NM_003683.5 | Refseq |
