Assay Details
| Gene Symbol: TTC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017931.2 | NP_060401.2 | 5 - 6 | 610 | 72 | |
| XM_011530259.1 | XP_011528561.1 | 6 - 7 | 738 | 72 | ||
| XM_011530260.2 | XP_011528562.1 | 5 - 6 | 610 | 72 | ||
| GenBank mRNA | AK000706.1 | - | 5 - 6 | 566 | 72 | |
| BC018918.2 | - | 5 - 6 | 551 | 4130906 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000393.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22007597.hg.1 | ENST00000381031.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007597.hg.1 | ENST00000445282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007597.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007597.hg.1 | TC22000393.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007595.hg.1 | ENST00000381031.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007595.hg.1 | ENST00000445282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007595.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007595.hg.1 | TC22000393.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000475467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000475467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000475467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000629918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000629918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | ENST00000629918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000417709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000451998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000475467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000475467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000475467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000629918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000629918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | ENST00000629918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007660.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | TC2200007660.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154440.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | NM_017931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | TC2200007660.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | ENST00000381031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | ENST00000421359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | ENST00000422713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200154436.hg.1 | NM_017931.2 | Refseq |

TTC38
Genomic Map