| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
ENST00000327628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
ENST00000341421.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
ENST00000396175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
ENST00000449246.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
ENST00000487159.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
ENST00000543338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
NM_001042646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
NM_014965.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004340.hg.1 |
TC03000217.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
ENST00000327628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
ENST00000341421.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
ENST00000396175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
ENST00000449246.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
ENST00000487159.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
ENST00000543338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
NM_001042646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
NM_014965.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004342.hg.1 |
TC03000217.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000327628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000341421.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000396175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000449246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000484786.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000613405.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001042646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001265608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001265609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001265610.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_014965.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000327628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000341421.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000396175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000449246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000484786.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
ENST00000613405.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001042646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001265608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001265609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
TC0300013812.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_014965.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203775.hg.1 |
NM_001265610.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000327628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000341421.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000396175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000427771.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000449246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000484786.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000613405.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001042646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001265608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001265609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001265610.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_014965.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000327628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000341421.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000396175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000427771.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000449246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000484786.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
ENST00000613405.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001042646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001265608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001265609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_001265610.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
NM_014965.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300203777.hg.1 |
TC0300013812.hg.1 |
Clariom D TC |