Assay Details
| Gene Symbol: USP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538800.2 | XP_011537102.1 | 2 - 3 | 246 | 133 | |
| XM_011538801.1 | XP_011537103.1 | 2 - 3 | 106 | 133 | ||
| XM_017020011.1 | XP_016875500.1 | 2 - 3 | 92 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538800.2, XM_011538801.1, XM_017020011.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic nuclear division
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
regulation of spindle checkpoint
regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of ubiquitin protein ligase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
zinc ion binding
mitotic nuclear division
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
regulation of spindle checkpoint
regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of ubiquitin protein ligase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000547951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000547951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000258499.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000393091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000537435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000547951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000547951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000549639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000551837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | ENST00000552440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001042403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_001278393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NM_032147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011588.hg.1 | NR_125360.2 | Refseq |

USP44
Genomic Map