| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
238093_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1566906_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022838.hg.1 |
ENST00000416962.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022838.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022838.hg.1 |
TC09001768.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022839.hg.1 |
ENST00000416962.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022839.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022839.hg.1 |
TC09001768.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022837.hg.1 |
ENST00000416962.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022837.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022837.hg.1 |
TC09001768.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022836.hg.1 |
ENST00000416962.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022836.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022836.hg.1 |
TC09001768.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900182966.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182966.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182966.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182966.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182966.hg.1 |
TC0900012049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900182970.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182970.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182970.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182970.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182970.hg.1 |
TC0900012049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900182971.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182971.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182971.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182971.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182971.hg.1 |
TC0900012049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900182967.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182967.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182967.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182967.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182967.hg.1 |
TC0900012049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900182969.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182969.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182969.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182969.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182969.hg.1 |
TC0900012049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012049.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182968.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182968.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182968.hg.1 |
ENST00000612170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182968.hg.1 |
NR_038389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182968.hg.1 |
TC0900012049.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
238093_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
238093_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238093_at4 |
- |
- |
| Human Genome U95 Set |
81676_PM_at |
- |
- |
| Human Genome U95 Set |
81676_at |
- |
- |
| Human Genome U133 Set |
238093_at |
- |
- |
| Human Genome U133 Set |
238093_PM_at |
- |
- |