Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014487.5 | NP_055302.1 | 2 - 3 | 349 | 105 | |
| XM_017008033.1 | XP_016863522.1 | 2 - 3 | 493 | 105 | ||
| GenBank mRNA | AJ006591.2 | - | 2 - 3 | 294 | 105 | |
| AK002106.1 | - | 2 - 3 | 299 | 105 | ||
| AK223609.1 | - | 2 - 3 | 300 | 105 | ||
| AK315463.1 | - | 2 - 3 | 335 | 105 | ||
| BC004421.1 | - | 2 - 3 | 325 | 3352916 | 105 | |
| DA089047.1 | - | 2 - 3 | 341 | 105 | ||
More Information
Set Membership: |
|

ZNF330
Genomic Map