Assay Details
| Gene Symbol: DDX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007204.4 | NP_009135.4 | 5 - 6 | 1177 | 88 | |
| GenBank mRNA | AF106019.1 | - | 5 - 6 | 830 | 88 | |
| AF155803.1 | - | 5 - 6 | 825 | 88 | ||
| AF171063.1 | - | 5 - 6 | 1113 | 88 | ||
| AK001506.1 | - | 5 - 6 | 839 | 88 | ||
| AK301697.1 | - | 4 - 5 | 678 | 88 | ||
| AK314312.1 | - | 5 - 6 | 822 | 88 | ||
| BC011556.1 | - | 5 - 6 | 830 | 4651697 | 88 | |
| BC031062.1 | - | 5 - 6 | 839 | 5273709 | 88 | |
| BC034953.2 | - | 5 - 6 | 1148 | 4824324 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000981.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
spliceosomal tri-snRNP complex assembly
spliceosomal snRNP assembly
RNA processing
negative regulation of cell proliferation
RNA secondary structure unwinding
positive regulation of apoptotic process
oogenesis
regulation of steroid biosynthetic process
nuclear import
DNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
protein domain specific binding
protein binding, bridging
histone deacetylase binding
repressing transcription factor binding
negative regulation of transcription from RNA polymerase II promoter
spliceosomal tri-snRNP complex assembly
spliceosomal snRNP assembly
RNA processing
negative regulation of cell proliferation
RNA secondary structure unwinding
positive regulation of apoptotic process
oogenesis
regulation of steroid biosynthetic process
nuclear import
DNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
protein domain specific binding
protein binding, bridging
histone deacetylase binding
repressing transcription factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01015657.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015657.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015657.hg.1 | ENST00000536167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015657.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015657.hg.1 | TC01000981.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167859.hg.1 | TC0100009428.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |

DDX20
Genomic Map