| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15015601.hg.1 |
ENST00000356056.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15015601.hg.1 |
ENST00000424560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15015601.hg.1 |
ENST00000444904.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15015601.hg.1 |
NM_006901.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15015601.hg.1 |
TC15001620.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000356056.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000444904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000561618.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000563071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000564571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
NM_006901.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
TC1500009946.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000561618.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000356056.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000444904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000563071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
ENST00000564571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173751.hg.1 |
NM_006901.3 |
Refseq |