| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015919.hg.1 |
ENST00000373275.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015919.hg.1 |
ENST00000473691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015919.hg.1 |
NM_153252.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015919.hg.1 |
TC0X001182.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
ENST00000373275.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
ENST00000473691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
NM_153252.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
ENST00000373275.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
ENST00000473691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
NM_153252.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00169368.hg.1 |
TC0X00010169.hg.1 |
Clariom D TC |