Assay Details
| Gene Symbol: BRWD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153252.4 | NP_694984.4 | 18 - 19 | 2307 | 140 | |
| XM_005262113.3 | XP_005262170.1 | 18 - 19 | 2309 | 140 | ||
| XM_017029384.1 | XP_016884873.1 | 14 - 15 | 1837 | 140 | ||
| XM_017029385.1 | XP_016884874.1 | 18 - 19 | 2309 | 140 | ||
| GenBank mRNA | AK095887.1 | - | 1 - 2 | 180 | 140 | |
| AY497046.1 | - | 18 - 19 | 2307 | 140 | ||
| AY497047.1 | - | 15 - 16 | 2239 | 140 | ||
| AY497048.1 | - | 18 - 19 | 2330 | 140 | ||
| AY497049.1 | - | 17 - 18 | 2300 | 140 | ||
| AY497050.1 | - | 18 - 19 | 2280 | 140 | ||
| AY497051.1 | - | 15 - 16 | 1867 | 140 | ||
| AY497052.1 | - | 20 - 21 | 2466 | 140 | ||
| AY497053.1 | - | 16 - 17 | 2055 | 140 | ||
| AY497054.1 | - | 15 - 16 | 1956 | 140 | ||
| AY497055.1 | - | 18 - 19 | 2241 | 140 | ||
| AY497056.1 | - | 15 - 16 | 2212 | 140 | ||
| AY497057.1 | - | 14 - 15 | 2111 | 140 | ||
| AY497058.1 | - | 14 - 15 | 1950 | 140 | ||
| AY497059.1 | - | 16 - 17 | 2321 | 140 | ||
| AY497060.1 | - | 12 - 13 | 1888 | 140 | ||
| BC111490.1 | - | 18 - 19 | 2257 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001182.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK095887.1 transcripts.
Set Membership: |
|

BRWD3
Genomic Map