Assay Details
| Gene Symbol: IFT172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015662.2 | NP_056477.1 | 21 - 22 | 2353 | 91 | |
| XM_005264254.1 | XP_005264311.1 | 21 - 22 | 2184 | 91 | ||
| XM_006711986.3 | XP_006712049.1 | 22 - 23 | 2445 | 91 | ||
| XM_006711987.1 | XP_006712050.1 | 21 - 22 | 2250 | 91 | ||
| XM_011532757.2 | XP_011531059.1 | 21 - 22 | 2559 | 91 | ||
| XM_011532758.1 | XP_011531060.1 | 21 - 22 | 2250 | 91 | ||
| XM_011532759.2 | XP_011531061.1 | 7 - 8 | 835 | 91 | ||
| XM_011532760.2 | XP_011531062.1 | 4 - 5 | 515 | 91 | ||
| XM_017003790.1 | XP_016859279.1 | 22 - 23 | 2379 | 91 | ||
| XM_017003791.1 | XP_016859280.1 | 20 - 21 | 2320 | 91 | ||
| XM_017003792.1 | XP_016859281.1 | 21 - 22 | 2250 | 91 | ||
| XM_017003793.1 | XP_016859282.1 | 17 - 18 | 1760 | 91 | ||
| XM_017003794.1 | XP_016859283.1 | 15 - 16 | 1463 | 91 | ||
| XM_017003795.1 | XP_016859284.1 | 19 - 20 | 2312 | 91 | ||
| GenBank mRNA | AB033005.1 | - | 3 - 4 | 218 | 91 | |
| AL110218.1 | - | 18 - 19 | 1909 | 91 | ||
| BC014849.1 | - | 6 - 7 | 598 | 91 | ||
| BC137126.1 | - | 21 - 22 | 2243 | 9021123 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

IFT172
Genomic Map