| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027275.hg.1 |
ENST00000260570.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027275.hg.1 |
NM_015662.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027275.hg.1 |
TC02001687.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000260570.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000450564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000507184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000509128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
NM_015662.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000260570.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000450564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000507184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
ENST00000509128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
NM_015662.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200188238.hg.1 |
TC0200012082.hg.1 |
Clariom D TC |