Assay Details
| Gene Symbol: MTCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317231.1 | NP_001304160.1 | 10 - 11 | 868 | 109 | |
| NM_001317232.1 | NP_001304161.1 | 9 - 10 | 841 | 109 | ||
| NM_001317233.1 | NP_001304162.1 | 9 - 10 | 810 | 109 | ||
| NM_014342.3 | NP_055157.1 | 10 - 11 | 868 | 109 | ||
| XM_011519959.2 | XP_011518261.1 | 10 - 11 | 811 | 109 | ||
| XM_011519960.2 | XP_011518262.1 | 9 - 10 | 831 | 109 | ||
| XM_017017463.1 | XP_016872952.1 | 10 - 11 | 787 | 109 | ||
| GenBank mRNA | AF085361.1 | - | 10 - 11 | 727 | 109 | |
| AF176008.1 | - | 10 - 11 | 728 | 109 | ||
| AK223097.1 | - | 10 - 11 | 691 | 109 | ||
| AK295218.1 | - | 9 - 10 | 690 | 109 | ||
| AK313032.1 | - | 10 - 11 | 811 | 109 | ||
| AY380792.1 | - | 10 - 11 | 726 | 109 | ||
| BC000875.1 | - | 10 - 11 | 724 | 3461469 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001746.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11020441.hg.1 | ENST00000302503.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020441.hg.1 | ENST00000534074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020441.hg.1 | ENST00000542981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020441.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020441.hg.1 | TC11001746.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008010.hg.1 | ENST00000562099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008010.hg.1 | ENST00000562099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020442.hg.1 | ENST00000302503.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020442.hg.1 | ENST00000534074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020442.hg.1 | ENST00000542981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020442.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020442.hg.1 | TC11001746.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000533571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | ENST00000539759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010757.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008010.hg.1 | ENST00000562099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008010.hg.1 | ENST00000562099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181575.hg.1 | TC1100010757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000302503.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000525649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000530428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | ENST00000534074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | NM_014342.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181574.hg.1 | TC1100010757.hg.1 | Clariom D TC |

MTCH2
Genomic Map