Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009716.hg.1 |
TC06000817.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009716.hg.1 |
ENST00000358812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009716.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009716.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161324.hg.1 |
TC0600008863.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000358812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000369293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000474553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
ENST00000612218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008863.hg.1 |
NM_024641.3 |
Refseq |