| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007188.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007188.hg.1 |
ENST00000426378.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007188.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007188.hg.1 |
TC08002581.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007186.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007186.hg.1 |
ENST00000426378.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007186.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007186.hg.1 |
TC08002581.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007185.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007185.hg.1 |
ENST00000426378.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007185.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007185.hg.1 |
TC08002581.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008124.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
ENST00000520921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155765.hg.1 |
TC0800008124.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
ENST00000285381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
ENST00000522207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
NM_005181.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155766.hg.1 |
TC0800008124.hg.1 |
Clariom D TC |