Assay Details
| Gene Symbol: CMPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256477.1 | NP_001243406.1 | 2 - 3 | 918 | 99 | |
| NM_001256478.1 | NP_001243407.1 | 2 - 3 | 918 | 99 | ||
| NM_207315.3 | NP_997198.2 | 2 - 3 | 918 | 99 | ||
| NR_046236.1 | 2 - 3 | 330 | 99 | |||
| GenBank mRNA | AK308509.1 | - | 3 - 4 | 442 | 99 | |
| BC089425.1 | - | 2 - 3 | 771 | 99 | ||
| BC132821.1 | - | 2 - 3 | 918 | 40146843 | 99 | |
| BC141802.1 | - | 2 - 3 | 811 | 40147165 | 99 | |
| BC144202.1 | - | 2 - 3 | 918 | 9052722 | 99 | |
| BX463706.2 | - | 2 - 3 | 226 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02005020.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleoside diphosphate phosphorylation
dUDP biosynthetic process
dTDP biosynthetic process
dTTP biosynthetic process
nucleoside triphosphate biosynthetic process
cellular response to lipopolysaccharide
cytidylate kinase activity
nucleoside diphosphate kinase activity
thymidylate kinase activity
ATP binding
UMP kinase activity
nucleoside diphosphate phosphorylation
dUDP biosynthetic process
dTDP biosynthetic process
dTTP biosynthetic process
nucleoside triphosphate biosynthetic process
cellular response to lipopolysaccharide
cytidylate kinase activity
nucleoside diphosphate kinase activity
thymidylate kinase activity
ATP binding
UMP kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024963.hg.1 | ENST00000256722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024963.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024963.hg.1 | ENST00000458098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024963.hg.1 | ENST00000478738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024963.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024963.hg.1 | TC02005020.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184454.hg.1 | TC0200011624.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184435.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184435.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184435.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184435.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184435.hg.1 | TC0200011624.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184446.hg.1 | TC0200011624.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184455.hg.1 | TC0200011624.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000256722.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000404168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000458098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000465619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000470479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000478738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | ENST00000491738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256477.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_001256478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NM_207315.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011624.hg.1 | NR_046236.1 | Refseq |

CMPK2
Genomic Map