Assay Details
| Gene Symbol: CNOT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017546.4 | NP_060016.3 | 4 - 5 | 1114 | 92 | |
| GenBank mRNA | AF103798.1 | - | 4 - 5 | 1105 | 92 | |
| AK024221.1 | - | 3 - 4 | 531 | 92 | ||
| BC018664.2 | - | 4 - 5 | 1023 | 92 | ||
| BC064421.1 | - | 4 - 5 | 1081 | 5752134 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000615.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA poly(A) tail shortening
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell proliferation
gene silencing by RNA
molecular_function
protein binding
nuclear-transcribed mRNA poly(A) tail shortening
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell proliferation
gene silencing by RNA
molecular_function
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02009696.hg.1 | ENST00000289382.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009696.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009696.hg.1 | TC02000615.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009698.hg.1 | ENST00000289382.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009698.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009698.hg.1 | TC02000615.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009697.hg.1 | ENST00000289382.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009697.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009697.hg.1 | TC02000615.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160313.hg.1 | TC0200008646.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008646.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | ENST00000462489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160312.hg.1 | TC0200008646.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | ENST00000289382.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | ENST00000420107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | NM_017546.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160314.hg.1 | TC0200008646.hg.1 | Clariom D TC |

CNOT11
Genomic Map