Assay Details
| Gene Symbol: DRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018370.2 | NP_060840.2 | 5 - 6 | 1039 | 97 | |
| XM_017019578.1 | XP_016875067.1 | 6 - 7 | 1161 | 97 | ||
| GenBank mRNA | AK002121.1 | - | 3 - 4 | 345 | 97 | |
| AK094923.1 | - | 5 - 6 | 669 | 97 | ||
| BC013773.1 | - | 3 - 4 | 337 | 97 | ||
| BC018435.1 | - | 5 - 6 | 1015 | 4474297 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000793.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010042.hg.1 | ENST00000258534.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010042.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010042.hg.1 | TC12000793.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163607.hg.1 | TC1200008629.hg.1 | Clariom D TC |

DRAM1
Genomic Map